Learn More
AdipoGen anti-Polyglutamylation Modification, mAb (GT335)
SDP
Catalog No. 50463394
Encompass_Preferred
anti-Polyglutamylation Modification, mAb (GT335), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes the posttranslational modification (poly)glutamylation on proteins. Reacts with polyglutamylated alpha- and beta-tubulin. Isotype: Mouse IgG1kappa. Clone: GT335. Applications: EM, ICC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Polyglutamylation is a post-translational modification in which glutamate side chains of variable lengths are formed on the modified protein. It is evolutionarily conserved and the most prominent substrate is tubulin, the microtubule (MT) building block. Polyglutamylation has been proposed to be involved in the functional adaptation of MTs, as it occurs within the carboxy-terminal tubulin tails that participate directly in the binding of many structural and motor MT-associated proteins.
Catalog No. 50-463-394 Supplier AdipoGen Supplier No. AG20B0020C100
anti-Polyglutamylation Modification, mAb (GT335), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes the posttranslational modification (poly)glutamylation on proteins. Reacts with polyglutamylated alpha- and beta-tubulin. Isotype: Mouse IgG1kappa. Clone: GT335. Applications: EM, ICC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Polyglutamylation is a post-translational modification in which glutamate side chains of variable lengths are formed on the modified protein. It is evolutionarily conserved and the most prominent substrate is tubulin, the microtubule (MT) building block. Polyglutamylation has been proposed to be involved in the functional adaptation of MTs, as it occurs within the carboxy-terminal tubulin tails that participate directly in the binding of many structural and motor MT-associated proteins.