Learn More
Immunology Consultants anti-Protein G Antibody (Chicken) - HRP Conjugated
SDP
Catalog No. 5000435307
Encompass_Preferred
This HRP‑conjugated chicken antibody enables direct, enzyme‑mediated detection of Protein G with strong chemiluminescent or colorimetric signal output. Ideal for ELISA and Western blot applications, it supports sensitive quantification of residual Protein G in purification workflows and biologics manufacturing. The avian origin reduces cross‑reactivity with mammalian proteins, ensuring low background and high assay clarity. Produced from affinity‑purified material, CPG‑65P offers consistent performance and is well suited for QC testing, impurity monitoring, and analytical methods requiring rapid, HRP‑based detection.
Catalog No. 50-004-35307 Supplier Immunology Consultants Supplier No. CPG65P
This HRP‑conjugated chicken antibody enables direct, enzyme‑mediated detection of Protein G with strong chemiluminescent or colorimetric signal output. Ideal for ELISA and Western blot applications, it supports sensitive quantification of residual Protein G in purification workflows and biologics manufacturing. The avian origin reduces cross‑reactivity with mammalian proteins, ensuring low background and high assay clarity. Produced from affinity‑purified material, CPG‑65P offers consistent performance and is well suited for QC testing, impurity monitoring, and analytical methods requiring rapid, HRP‑based detection.