Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Prostate Specific Antigen (PSA), 0.2mg
Supplier: Advanced ImmunoChemical, Inc. 2PSA0.2MG
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal Antibodies/Tumor Markers/Monoclonal mouse anti-prostate specific antigen (PSA). MAbs:2H9, 5A6, 8A6, 5G6, PS1, PS2, 1H12; Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human PSA antigen purified from seminal plasma. MAb Isotypes:IgG1 for MAbs 5G6, PS2, PS1; IgG2a for MAbs 2H9, 5A6, 8A6, 1H12; Detection of PSA antigen. Recommended pairs for detection of total PSA (capture-detection): 5A6-2H9, 5G6 – 5A6. Best pair: 1H12 – 5A6; Recommended pairs for detection of free PSA (capture-detection): 5A6-8A6, 2H9-8A6, PS2-PS1, PS1-PS2. Best pair: 8A6 – 5A6; MAbs can also be used in lateral flow. MAbs PS1 and PS2 by ion exchange chromatography & other Abs by chromatography on protein A Sepharose; MAbs 2H9, 5A6, 8A6, 5G6 and 1H12 in PBS, pH 7.4, 0.1 % sodium azide (NaN3) MAbs PS1 and PS2 in 0.15 M NaCl.