Learn More
ECM Bioscience PYK2 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM2291
Encompass_Preferred
Clone M229 was generated from a recombinant human PYK2 protein that included amino acids residues in the C-terminal region. This sequence is conserved in rat and mouse PYK2, and has low homology to other related kinases. Accession #Q14289. The antibody detects a 116 kDa* band corresponding to PYK2 on SDS-PAGE immunoblots of mouse brain tissue. Applications: WB, E, ICC; Species Reactivity: Hu, Rt, Ms; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = CAKbeta, FAK2
Catalog No. 50-206-8793