Learn More
BIOHIPPO Anti-Pyrroline-5-carboxylate/P5C Monoclonal antibody (1A055), 100 µg
SDP
Catalog No. 5000858331
Encompass_Preferred
General Pyrroline-5-carboxylate/P5C Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes Pyrroline-5-carboxylate/P5C. It shows reactivity with General. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.
Catalog No. 50-008-58331 Supplier BIOHIPPO Supplier No. BHA21416321
General Pyrroline-5-carboxylate/P5C Monoclonal Antibody is a Mouse-derived monoclonal antibody that specifically recognizes Pyrroline-5-carboxylate/P5C. It shows reactivity with General. Validated applications include ELISA, IHC, WB. Protein A/G purified from cell culture supernatant. Supplied as 100 ug.