Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Goat Anti-Rabbit IgG, Fc fragment specific, 1.5ml
SDP
Catalog No. NC9448271
Encompass_Preferred
Peroxidase-AffiniPure Goat Anti-Rabbit IgG, Fc Fragment Specific (min X Hu Sr Prot)
Catalog No. NC9448271 Supplier Jackson Immuno Research Labs Supplier No. 111035046
Peroxidase-AffiniPure Goat Anti-Rabbit IgG, Fc Fragment Specific (min X Hu Sr Prot)