Learn More
Sigma Aldrich Fine Chemicals Biosciences Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, MFCD00162782, 0.25mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A3687.25ML
Encompass
Affinity isolated antibody, buffered aqueous glycerol solution, Synonym: Alkaline Phosphatase Secondary Antibody, Anti Rabbit Antibody - Anti-Rabbit IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat, Anti Rabbit Sigma, anti rabbit antibody; IgGs are glycoprotein antibodies that modulate several immune responses. Rabbit IgGs against target proteins are often used as primary antibodies in various research applications. Thus, secondary anti-rabbit IgGs conjugated to a detectable substrate are useful tools for the analysis of target proteins. Goat anti-Rabbit IgG (whole molecule)-Alkaline Phosphatase antibody binds all rabbit Igs.
Catalog No. NC1430935