Learn More
Vector Laboratories Anti-DNP, Unconjugated, 1 mg DI-1088-1.5
SDP
Catalog No. 502045635
Encompass
Anti-DNP (Dinitrophenyl), Unconjugated, 1 mg. Dinitrophenyl (DNP) is not found endogenously in tissues and is an excellent alternative to biotin for use in applications requiring biotin-free systems. Probes or antibodies labeled with DNP can be detected using alkaline phosphatase–labeled DNP or using the biotinylated antibody followed by a streptavidin or avidin system.
Catalog No. 50-204-5635 Supplier Vector Laboratories Supplier No. DI10881.5
Anti-DNP (Dinitrophenyl), Unconjugated, 1 mg. Dinitrophenyl (DNP) is not found endogenously in tissues and is an excellent alternative to biotin for use in applications requiring biotin-free systems. Probes or antibodies labeled with DNP can be detected using alkaline phosphatase–labeled DNP or using the biotinylated antibody followed by a streptavidin or avidin system.