Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB AP
Supplier:  Seracare Life Sciences Inc 52200308
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-670-08