Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB DL680
Supplier:  Seracare Life Sciences Inc 52300402
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from goats immunized with purified rabbit IgG was labeled with Dylight 680 using a proprietary method. DyLight conjugates provide increased brightness and greater photostability than other fluorophores (ex. -- FITC). This conjugate is designed for use in immunofluorescense assays, including flow cytometry, immunohistology, Western blotting, microarrays and FLISA.
Catalog No. 50-671-40