Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB FITC
Supplier:  Seracare Life Sciences Inc 52300298
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product is in lyophilized form.
Catalog No. 50-668-84