Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200336
Encompass_Preferred
Anti-Rabbit IgG (H+L) Antibody, Peroxidase labeled. 1 mg. Affinity purified polyclonal antibody to Rabbit IgG heavy and light chains (H+L) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-00