Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 54500010
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-87