Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB HRP
Supplier:  Seracare Life Sciences Inc 52200283
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
Catalog No. 50-669-38