Learn More
Seracare Life Sciences Inc ANTI-RABBIT IGG (H+L) AB HSA
Supplier:  Seracare Life Sciences Inc 52100176
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, both heavy chain and light chains (whole IgG) made in Goat. Product has been cross-adsorbed with human serum to reduce cross-reactivity to human immunoglobulin.
Catalog No. 50-668-17