Learn More
Immunology Consultants Affinity Purified Donkey anti-Rabbit IgG H+L Antibody - Highly Cross-Adsorbed, 1.0 mg
SDP
Catalog No. 502414306
Encompass_Preferred
Donkeys were immunized with highly purified rabbit IgG h+l and resulting antiserum was collected. Antibody was solid phase adsorbed using bovine, chicken, goat, guinea pig, horse, hamster, human, mouse, rat and sheep immunosorbents to remove cross reactive antibody. Specific antibody was immunoaffinity purified off a rabbit IgG containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody will react with rabbit IgG and with light chains common with other rabbit immunoglobulins as determined by ELISA and IEP techniques. It has less than 1% cross reactivity with bovine, goat, guinea pig, horse, hamster, human, mouse, rat and sheep IgG and chicken IgY. This antibody is suitable for blotting, ELISA and IHC applications. This antibody may cross react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4306 Supplier Immunology Consultants Supplier No. DGHL15AXSPP
Donkeys were immunized with highly purified rabbit IgG h+l and resulting antiserum was collected. Antibody was solid phase adsorbed using bovine, chicken, goat, guinea pig, horse, hamster, human, mouse, rat and sheep immunosorbents to remove cross reactive antibody. Specific antibody was immunoaffinity purified off a rabbit IgG containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody will react with rabbit IgG and with light chains common with other rabbit immunoglobulins as determined by ELISA and IEP techniques. It has less than 1% cross reactivity with bovine, goat, guinea pig, horse, hamster, human, mouse, rat and sheep IgG and chicken IgY. This antibody is suitable for blotting, ELISA and IHC applications. This antibody may cross react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.