Learn More
Rockland Immunochemicals Anti-RABBIT IgG (H&L) (DONKEY) Antibody DyLight™ 800 Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Ms Rt & Sh Serum Proteins) - 611-745-127
SDP
Catalog No. 501058595
Encompass_Preferred
Anti-Rabbit IgG (H&L) DyLight 800 Antibody is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8595 Supplier Rockland Immunochemicals Supplier No. 611745127
Anti-Rabbit IgG (H&L) DyLight 800 Antibody is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.