Learn More
Rockland Immunochemicals Anti-RABBIT IgG (H&L) (GOAT) Antibody DyLight™ 680 Conjugated - 611-144-002
SDP
Supplier:  Rockland Immunochemicals 611144002
Encompass
Anti-Rabbit IgG Antibody DyLight™680 generated in goat detects rabbit IgG. This antibody will react with heavy chains of Rabbit IgG and with light chains of most Rabbit immunoglobulins. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Rabbit IgG (H&L) is conjugated to DyLight™680. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8547