Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Supplier:  Rockland Immunochemicals 6116302
Encompass
Anti-Rabbit IgG peroxidase conjugated antibody generated in sheep detects specifically rabbit IgG. This secondary peroxidase conjugated antibody anti-Rabbit is ideal for investigators who routinely perform titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3927