Learn More
Rockland Immunochemicals Rabbit IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943928
Encompass_Preferred
Anti-Rabbit IgG (H&L) peroxidase Antibody generated in donkey detects reactivity to Rabbit IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3928 Supplier Rockland Immunochemicals Supplier No. 6117302
Anti-Rabbit IgG (H&L) peroxidase Antibody generated in donkey detects reactivity to Rabbit IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.