Learn More
Rockland Immunochemicals Anti-RABBIT IgG (H&L) (SHEEP) Antibody DyLight™ 488 Conjugated (Min X Bv Ch Gt GP Hs Hu Ms Rt & Sh Serum Proteins) - 611-641-122
SDP
Supplier:  Rockland Immunochemicals 611641122
Encompass
Anti-Rabbit IgG Antibody DyLight™488 generated in sheep detects rabbit IgG. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat or Sheep Serum Proteins.  This antibody will react with heavy chains of rabbit IgG and with light chains of most rabbit immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This Anti-Rabbit IgG (H&L) is conjugated to DyLight™488. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.
Catalog No. 50-105-8575