Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Anti-rabbit IgG, HRP-linked Antibody 5 ml
SDP

Catalog No. NC0470879
Encompass

Anti-rabbit IgG, HRP-linked Antibody 5 ml

Catalog No. NC0470879 Supplier Cell Signaling Technology Supplier No. 7074V
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anti-rabbit IgG, HRP-linked Antibody 5 ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.