Learn More
Vector Laboratories Goat Anti-Rabbit IgG Antibody Texas Red 1.5 mg
SDP
Supplier:  Vector Laboratories TI10001.5
Encompass
Texas Red Goat Anti-Rabbit IgG secondary antibodies are prepared by hyperimmunizing animals to produce high-affinity antibodies, and purifying the antibodies by affinity chromatography. This Texas Red conjugate is prepared in a manner that ensures the maximum degree of labeling with no alteration in the specificity or affinity of the antibody. Texas Red (sulforhodamine 101) excites at about 595 nm and emits at about 615 nm, producing a bright red fluorescence well removed from the emission spectrum of fluorescein. This conjugate is ideal as a single or double label reagent for flow cytometry or tissue staining.
Catalog No. NC9844418