Learn More
Abcam VeriBlot for IP Detection Reagent (HRP), 500UL
Supplier:  Abcam AB131366500UL
Encompass_Preferred
VeriBlot for IP detection reagent (HRP) is an immunoblotting reagent that enables the trouble-free detection of immunoblotted target proteins without interference from denatured IgG. - Cited in over 80 publications - Detects target proteins without interference from IgG heavy and light chains - Recognizes non-reduced antibodies, minimizing detection of denatured IgG - Multiple species compatibility – detects IgG from bovine, goat, human, mouse, rat, rabbit, and sheep (specific isotypes detailed) - Suitable for up to 20 blots with proper dilution (1:200 in 5 ml milk)
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0466233