Learn More
Echelon Biosciences Research Labs ANTI-RAT ATP SYNTHASE 0.1 mL
SDP
Supplier:  Echelon Biosciences Research Labs ZRATPA100UL
Encompass
ATP synthase is comprised of 2 major units, the Fo and F1. Five different subunits make up the F1 unit: 3 , 3 , 1 , 1 , and 1 subunits. The and subunit are localized to the mitochondrial and plasma membranes and the subunit is thought to be involved in the satiation response through binding to enterostatin. This item is non returnable
Catalog No. 50-720-933