Learn More
AbD Serotec ANTI RAT HUMAN CD52:FITC
Supplier:  AbD Serotec MCA1642F
Encompass
Rat Anti Human, Specificity: CD52, Product Type: Monoclonal Antibody, Clone: YTH34.5, Isotype: IgG2b, Quantity/Volume: 0.1mg, Form: Purified IgG conjugated to Fluorescein Isothiocyanate Isomer 1 (FITC) - liquid
Catalog No. NC9144070