Learn More
Immunology Consultants Affinity Purified Goat anti-Rat IgE Antibody, 0.5 mg
SDP
Supplier:  Immunology Consultants GE25A
Encompass_Preferred
Goats were immunized with highly purified rat IgE and the resulting antiserum was collected. Antibody was solid phase adsorbed then immunoaffinity purified off a rat IgE containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with rat IgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may cross react with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4327