Learn More
Immunology Consultants HRP Conjugated Goat anti-Rat IgE Antibody, 0.5 mg
SDP
Catalog No. 502414286
Encompass_Preferred
Goats were immunized with highly purified rat IgE and the resulting antiserum was collected. Antibody was solid phase adsorbed, immunoaffinity purified off a rat IgE containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with rat IgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may crossreact with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4286 Supplier Immunology Consultants Supplier No. GE25P
Goats were immunized with highly purified rat IgE and the resulting antiserum was collected. Antibody was solid phase adsorbed, immunoaffinity purified off a rat IgE containing immunosorbent and conjugated with horseradish peroxidase (HRP). Antibody concentration was determined prior to conjugation using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with rat IgE as determined by IEP and ELISA techniques. It is suitable for blotting, ELISA and IHC applications. This antibody may crossreact with IgE from other species. Optimal working dilutions should be determined experimentally by the investigator.