Learn More
Vector Laboratories Rabbit Anti-Rat IgG Antibody Mouse Adsorbed Unconjugated 0.5 mg
SDP
Catalog No. NC9885522
Encompass
Unconjugated Rabbit Anti-Rat IgG antibodies are purified by an affinity chromatography procedure to remove any low affinity antibodies, then cross-reactive antibodies are removed by solid-phase adsorption. The final product is subjected to rigorous quality control assays including immunodiffusion, solid-phase enzyme immunoassays, gel electrophoresis and solid-phase binding assays. These antibodies will recognize both heavy and light chains. Rabbit Anti-Rat IgG Antibody is supplied in solution. This ultrapure, high affinity antibody is ready for iodination, fluorochrome labeling, or enzyme conjugations. It can also be employed as a capture antibody in enzyme immunoassays or in other assays requiring carrier-free immunoglobulins.
Catalog No. NC9885522 Supplier Vector Laboratories Supplier No. AI4001.5
Unconjugated Rabbit Anti-Rat IgG antibodies are purified by an affinity chromatography procedure to remove any low affinity antibodies, then cross-reactive antibodies are removed by solid-phase adsorption. The final product is subjected to rigorous quality control assays including immunodiffusion, solid-phase enzyme immunoassays, gel electrophoresis and solid-phase binding assays. These antibodies will recognize both heavy and light chains. Rabbit Anti-Rat IgG Antibody is supplied in solution. This ultrapure, high affinity antibody is ready for iodination, fluorochrome labeling, or enzyme conjugations. It can also be employed as a capture antibody in enzyme immunoassays or in other assays requiring carrier-free immunoglobulins.