Learn More
Seracare Life Sciences Inc ANTI-RAT IGG (GAMMA) AB AP
Catalog No. 5067009
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Rat IgG, heavy chains (gamma) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-670-09 Supplier Seracare Life Sciences Inc Supplier No. 52200309
Affinity purified polyclonal secondary antibody against Rat IgG, heavy chains (gamma) made in Goat and labeled with alkaline phosphatase. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.