Learn More
Seracare Life Sciences Inc ANTI-RAT IGG (H+L) AB CY3
Supplier:  Seracare Life Sciences Inc 52300361
Encompass_Preferred
Affinity purified polyclonal antibody to Rat IgG, both heavy and light chains (whole IgG) made in Goat and labeled with Cy3. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-43