Learn More
Seracare Life Sciences Inc ANTI-RAT IGG (H+L) AB DL680
Catalog No. 5067142
Encompass_Preferred
Affinity purified polyclonal antibody to rat IgG, both heavy and light chains (whole IgG) made in goat and labeled with DyLight 680. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-671-42 Supplier Seracare Life Sciences Inc Supplier No. 52300404
Affinity purified polyclonal antibody to rat IgG, both heavy and light chains (whole IgG) made in goat and labeled with DyLight 680. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).