Learn More
Seracare Life Sciences Inc ANTI-RAT IGG(H+L)AB MSA BIOTIN
Supplier:  Seracare Life Sciences Inc 52600043
Encompass
Affinity purified polyclonal antibody to Rat IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product has been cross adsorbed to mouse immunoglobulin to minimize cross reactivity. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-71