Learn More
Seracare Life Sciences Inc ANTI-RAT IGG (H+L) AB MSA HRP
Supplier:  Seracare Life Sciences Inc 52200365
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Rat IgG, both heavy and light chains (whole IgG) made in Goat and labeled with horseradish peroxidase. Product has been cross-adsorbed with mouse immunoglobulin to reduce cross-reactivity. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-27