Learn More
Seracare Life Sciences Inc ANTI-RAT IGG (H+L) ANTIBODY
Supplier:  Seracare Life Sciences Inc 52100179
Encompass_Preferred
Affinity purified polyclonal antibody to Rat IgG, both heavy chain and light chains (whole IgG) made in Goat. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-668-20