Learn More
Rockland Immunochemicals Rat IgG (H&L) Antibody Peroxidase Conjugated Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 612103120
Encompass
Anti-Rat IgG whole molecule antibody generated in goat detects specifically Rat IgG whole molecule. This secondary antibody anti-Rat is ideal for investigators who routinely perform ELISA Sandwich ELISA titration assays western-blot immunoprecipitation and more generally immunoassays.
Catalog No. 50-194-3930