Learn More
Rockland Immunochemicals Rat IgG (H&L) Secondary Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943934
Encompass_Preferred
HRP secondary antibody conjugated Rat IgG is Suitable for western blot ELISA immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Catalog No. 50-194-3934 Supplier Rockland Immunochemicals Supplier No. 6124302
HRP secondary antibody conjugated Rat IgG is Suitable for western blot ELISA immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.