Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Rat IgG2a, lambda, Clone: B39-4, BD

Catalog No. BDB563394 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563394 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563394 Supplier BD Biosciences Supplier No. 563394
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Host Species: Rat
Clone: B39-4
Isotype: IgG2a, lambda
Species Reactivity: Ms
Immunogen: Trinitrophenyl-Keyhole Limpet Hemocyanin

Isotype Control

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat IgG2a, lambda
Classification Monoclonal
Clone B39-4
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Host Species Rat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.