Learn More
Seracare Life Sciences Inc ANTI-RAT IGM (MU) AB HRP
Catalog No. 5066940
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Rat IgM, heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
This item is not returnable. View return policy
Catalog No. 50-669-40 Supplier Seracare Life Sciences Inc Supplier No. 52200285
Affinity purified polyclonal secondary antibody against Rat IgM, heavy chains (mu) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.