Learn More
Medchemexpress LLC Anti-Rat Kappa Immun 5mg | 5mg
SDP
Catalog No. 5000324668
Encompass_Preferred
Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) is an anti-rat Kappa Immunoglobulin Light Chain IgG2a monoclonal antibody Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can enhance B cell depletion Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can enhance T cell exhaustion after the injection of CD4 mAb (HY-P990792) and CD8 mAb (HY-P99129) Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can be used for research on immunology[1][2]
Catalog No. 50-003-24668 Supplier Medchemexpress LLC Supplier No. HYP9901205MG
Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) is an anti-rat Kappa Immunoglobulin Light Chain IgG2a monoclonal antibody Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can enhance B cell depletion Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can enhance T cell exhaustion after the injection of CD4 mAb (HY-P990792) and CD8 mAb (HY-P99129) Anti-Rat Kappa Immunoglobulin Light Chain Antibody (MAR 18 5) can be used for research on immunology[1][2]