Learn More
FUJIFILM Biosciences Anti Rat LC3 Rabbit 50uL
Catalog No. 5000450136
Encompass_Preferred
LC3 is a mammalian analogue of Atg8 which is a factor required for autophagy in budding yeast It is synthesized in the cytoplasm and immediately undergoes C-terminal cleavage to form LC3-I LC3-I then reacts with E1-like enzyme Atg7 and E2-like enzyme Atg3 and binds with phospholipids to form LC3-II LC3-II then binds to autophagosome membranes This product is a rabbit polyclonal antibody that recognizes LC3 Specificity to LC3
Catalog No. 50-004-50136 Supplier FUJIFILM Biosciences Supplier No. 01022841
LC3 is a mammalian analogue of Atg8 which is a factor required for autophagy in budding yeast It is synthesized in the cytoplasm and immediately undergoes C-terminal cleavage to form LC3-I LC3-I then reacts with E1-like enzyme Atg7 and E2-like enzyme Atg3 and binds with phospholipids to form LC3-II LC3-II then binds to autophagosome membranes This product is a rabbit polyclonal antibody that recognizes LC3 Specificity to LC3