Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) AB BIOTIN
Supplier:  Seracare Life Sciences Inc 52600038
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, heavy and light chains (whole IgG) made in Goat and biotinylated. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-672-66