Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) AB BIOTIN
Supplier:  Seracare Life Sciences Inc 55700007
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG heavy and light chains (whole IgG) made in Goat and biotinylated. This product is specifically for use in immunohistochemistry assays and is provided as a liquid at use dilution. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-675-72