Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) AB DL800
Supplier:  Seracare Life Sciences Inc 52300412
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from goats immunized with purified rabbit IgG was labeled with Dylight 800 using a proprietary method. DyLight conjugates provide increased brightness and greater photostability than other fluorophores (ex. -- FITC). This conjugate is designed for use in immunofluorescense assays, including flow cytometry, immunohistology, Western blotting, microarrays and FLISA.
Catalog No. 50-671-54