Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) AB HSA FITC
Catalog No. 5066885
Encompass_Preferred
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross- adsorbed to human serum to minimize cross-reactivity.
This item is not returnable. View return policy
Catalog No. 50-668-85 Supplier Seracare Life Sciences Inc Supplier No. 52300299
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC). Product has been cross- adsorbed to human serum to minimize cross-reactivity.