Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) AB XSA HRP
Supplier:  Seracare Life Sciences Inc 52200335
Encompass_Preferred
Affinity purified polyclonal antibody to rabbit IgG, heavy and light chains (whole Igg) made in goat and labeled with horseradish peroxidase. Product has been cross adsorbed to bovine, chicken, goat, guinea pig, hamster, horse, human, rabbit, rat and sheep to minimize cross-reactivity to these species. Product is in lyophilized form.
Catalog No. 50-672-01