Learn More
Seracare Life Sciences Inc ANTI-RB IGG (H+L) F(AB )2 HRP
Catalog No. 5067364
Encompass_Preferred
Affinity purified polyclonal antibody, F(ab)2 fragment, to Rabbit IgG, heavy and light chains (whole IgG). Made in Goat and labeled with horseradish peroxidase. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
This item is not returnable. View return policy
Catalog No. 50-673-64 Supplier Seracare Life Sciences Inc Supplier No. 52200393
Affinity purified polyclonal antibody, F(ab)2 fragment, to Rabbit IgG, heavy and light chains (whole IgG). Made in Goat and labeled with horseradish peroxidase. Product has been cross adsorbed to human serum to minimize cross reactivity to human immunoglobulin. Product is in lyophilized form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.