Learn More
Seracare Life Sciences Inc ANTI-RBIGG(H+L)F(AB )2HSADL488
Catalog No. 5067325
Encompass_Preferred
Affinity purified F(ab')2 fragment polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
This item is not returnable. View return policy
Catalog No. 50-673-25 Supplier Seracare Life Sciences Inc Supplier No. 52300430
Affinity purified F(ab')2 fragment polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).