Learn More
Seracare Life Sciences Inc ANTI-RBIGG(H+L)F(AB )2HSADL680
Supplier:  Seracare Life Sciences Inc 52300432
Encompass_Preferred
Affinity purified polyclonal F(ab'2) antibody fragment to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 680. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-673-41