Learn More
Trilink Bio Technologies Inc ARCA, 1 umole
SDP
Catalog No. NC1271217
Encompass
A key step in cellular mRNA processing is the addition of a 5’ cap structure, which is a 5'-5' triphosphate linkage between the 5' end of the RNA and a guanosine nucleotide. The cap is methylated enzymatically at the N-7 position of the guanosine to form mature mCAP. When preparing synthetic mRNA, the cap is often added prior to use in order to stabilize the mRNA and significantly enhance translation. Using a 4:1 mixture of a cap analog to GTP in transcription reactions will cap 80% of the resulting mRNAs. If mCAP (pre-methylated CAP) is used, only half of the asymmetrical mCAP will insert in the functional orientation. ARCA can only insert in the correct orientation, resulting in mRNAs that are translated twice as efficiently.
Catalog No. NC1271217 Supplier Trilink Bio Technologies Inc Supplier No. N70031
A key step in cellular mRNA processing is the addition of a 5’ cap structure, which is a 5'-5' triphosphate linkage between the 5' end of the RNA and a guanosine nucleotide. The cap is methylated enzymatically at the N-7 position of the guanosine to form mature mCAP. When preparing synthetic mRNA, the cap is often added prior to use in order to stabilize the mRNA and significantly enhance translation. Using a 4:1 mixture of a cap analog to GTP in transcription reactions will cap 80% of the resulting mRNAs. If mCAP (pre-methylated CAP) is used, only half of the asymmetrical mCAP will insert in the functional orientation. ARCA can only insert in the correct orientation, resulting in mRNAs that are translated twice as efficiently.